Imprimer
Stoppers, rubber, conical, without holes
Fournisseur: DEUTSCH NEUMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1010147 1010150 1010156 1010160 1010164 1010171 1010179 1010187 1010194 1010150/12/20
217-9416EA 8.5 EUR
217-9416 217-9417 217-9418 217-9419 217-9420 217-9421 217-9422 217-9423 217-9424 217-0823
Stoppers, rubber, conical, without holes
Stoppers Rubber Stoppers
Conical natural grey rubber stoppers without holes.
  • Excellent sealing and good adhesion
  • Good chemical resistance
  • Convenient pack sizes
  • Temperature resistant from −25 °C to +70 °C (+120 °C in humid atmosphere)
  • Shore hardness: 40 °A
Elastic, floats in water, non toxic.
Certifications: DIN 12871