Imprimer
Tubing valves
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8605-0100 8605-0060 8605-0080 8606-0100 8606-0060 8606-0080 8607-0100 8607-0060 8607-0080 8608-0100 8608-0060 8608-0080 8608-0120 8606-0120 8605-0120 8607-0120
229-7426EA 14.2 EUR
229-7426 229-7424 229-7425 229-7432 229-7430 229-7431 229-7423 229-7421 229-7422 229-7429 229-7427 229-7428 229-0115 229-0113 229-0112 229-0114
Tubing valves
Stopcocks
PE/PP, white and red or PVDF, translucent.
  • PVDF, excellent chemical resistance, suitable for use with foodstuffs
  • PP/PE for industrial applications
  • Maximum pressure load 1 bar
These two or three-way valves are ideally designed to regulate or completely stop the flow of gases and liquids. PVDF or PP/PE are preferred materials, particularly in the chemical, biological, pharmaceutical and food industries. Supplied with tube couplings to secure tube firmly.