Imprimer
VWR®, Barreaux d'agitation magnétiques, micro
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
442-0359EA 48.1 EUR
442-0359 442-0360 442-0361 442-0362 442-0363 442-0364 442-0365 442-0366 442-0367 442-0368 442-0447 442-0448 442-0449 442-0450 442-0451 442-0452 442-0453 442-0454 442-0455 442-0456 442-0457 442-0458 442-0459 442-0460 442-0461 442-0462 442-0463 442-0464 442-0465 442-0466 442-0467 442-0468 442-0469 442-0470 442-0471 442-0472 442-0473 442-0594 442-0595 442-0596 442-0597
VWR®, Barreaux d'agitation magnétiques, micro
Stirrers Magnetic Stirring Bars
PTFE covered, strong Alnico V magnetic core. Round, smooth surface. For use in the smallest containers.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)