Imprimer
Splash heads, vertical distillation, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH7/12 SH7/13 SH7/53
201-0260EA 126 EUR
201-0260 201-0261 201-0262
Splash heads, vertical distillation, Quickfit®
Composants de distillation Têtes de distillation
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
A splash head for use in distillation assemblies in which the condenser is assembled in the vertical mode. Lower cone accepts flask. Upper cone accepts condenser. Splash head reduces entrainment of raw liquid into the vapour stream.