Imprimer
Adapters, still heads, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH4/22 SH4/33 SH4/44 SH4/2 SH4/1 SH4/23
201-8696EA 54.9 EUR
201-8696 201-0616 201-8701 201-0614 201-1078 201-0757
Adapters, still heads, Quickfit®
Distillation Components Distillation Heads
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Used in distillation assemblies for connecting flasks to condenser. Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket.