Imprimer
Laboratory spoon with spatula end
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3392 3391 3393 3394 3390
231-1065EA 9.4 EUR
231-1065 231-1064 231-1066 231-1067 231-1063
Laboratory spoon with spatula end
Cuillères
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean