Imprimer
Pharmacist spoons
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3360 3370 3380 3385
BOCH3360EA 4.3 EUR
BOCH3360 231-1281 231-1282 BOCH3385
Pharmacist spoons
Cuillères
Stainless steel 18/10, anti-magnetic. Multipurpose spoons for a wide range of uses.
  • Narrow shape with extra long, flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean