Imprimer
Lens cleaning tissues, grade 105
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2105-862 2105-918 2105-841
111-5001EA 136 EUR
111-5001 111-5002 111-5000
Lens cleaning tissues, grade 105
Wipes Disposable Wipes and Towels Lens Papers
These cleaning tissues are suitable for use with lenses and any other optical surfaces made from glass, quartz or plastic.
  • Soft surface
  • Strong and highly absorbent
  • Chemically pure tissue free from silicones and other additives