Imprimer
Scoops
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4020 4021 4022 4023
231-0359EA 119 EUR
231-0359 NICK4021 NICK4022 NICK4023
Scoops
Pelles
Stainless steel. Round bottom. Ideal for use as weighing boats etc.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning