Imprimer
Feet for rods, Labomeca®
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
241-0137EA 359 EUR
241-0137 241-0136
Feet for rods, Labomeca®
Retort Stands
Mounting brackets for Labomeca® assemblies.
  • Used for attaching rods to a fixed point of the laboratory: floor, walls, ceiling, tables, shelves and furniture
  • Supplied with wood screw, can be secured by using Labomeca® Allen key
Informations de livraison: Supplied with wood screw.