Imprimer
Reaction vessels, round bottom, with flat flange, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FR1LF FR250F FR500F FR2LF FR5LF FR10LF FR20LF FR3LF FR700F
201-8495EA 138 EUR
201-8495 201-8511 201-8545 201-8521 201-8550 201-8478 201-8496 201-8527 BIBBFR700F
Reaction vessels, round bottom, with flat flange, Quickfit®
Récipients de réaction Flacons à réactions
PYREX®, borosilicate glass.
  • For use with Quickfit® flat flange lids and retaining clips
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.