Imprimer
Submersible plastic test tube grip racks, Bel-Art No-Wire, SP Bel-Art
Fournisseur: Bel-Art
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
No-Wire™
212-6265EA 82.8 EUR
212-6265 212-6267 212-6266
Submersible plastic test tube grip racks, Bel-Art No-Wire, SP Bel-Art
Portoirs Tube Racks
Retainer fingers hold tubes firmly in the rack plates with no fear of dislodging, making them perfect for both decanting and water bath work.
Tube size indicated is for grip function, smaller tubes may fit for storage purposes.