Imprimer
Porte-tubes à essai, No-Wire™, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F187450002 F187450004 F187450003 F187460002 F187460003 F187460004 F187470001 F187470003 F187470004 F187450001 F187460000 F187470002 F187450000 F187460001 F187470000
212-0582EA 68 EUR
212-0582 212-6581 212-6580 212-6582 212-6583 212-6584 212-6585 212-6586 212-6587 212-0579 212-0092 212-0581 212-0580 212-0584 212-0583
Porte-tubes à essai, No-Wire™, SP Bel-Art
Portoirs Tube Racks
PP, coloured.
  • Also perfect for use outside the water bath
  • Steam autoclavable at 121 °C
The economical alternative to wire racks. Due to their special blend of materials, these racks can be submerged without additional weight on the base of water baths and can also be used in shaking water baths. They have alphanumeric position markings for better sample identification and can be labelled on the side. Stackable for space saving storage.