Imprimer
Glass density bottles with thermometer, adjusted
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05640013 05640022 05640028 05640037
641-0903EA 270 EUR
641-0903 641-0904 641-0902 641-0901
Glass density bottles with thermometer, adjusted
Flacons de gravité spécifique
Borosilicate glass 3.3. Duran®, DIN 12809, ISO 3507, Gay-Lussac type.
  • Thermometer range of 10 to 35 °C
  • Thermometer has 0,2 °C divisions
  • 100 cm³ bottle is not according to DIN
The density bottles (pyknometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³. The bottle has a side arm with capillary with conical ground glass cap. The toluene thermometer has a frosted, enclosed glass scale.