Imprimer
Density bottles, adjusted
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05630210 05630213 05630222 05630228 05630237
LENZ05630210EA 87.9 EUR
LENZ05630210 LENZ05630213 LENZ05630222 LENZ05630228 641-0900
Density bottles, adjusted
Flacons de gravité spécifique
Borosilicate glass 3.3. DURAN®, Gay-Lussac type.
  • The density bottles (pycnometers) are individually engraved with the actual capacity, with an accuracy of 0,001 cm³
  • The top of stopper is ground and polished for ease of wiping clean
  • Stopper is size NS 10/19 with capillary
* DIN ISO 3507