Imprimer
Pointes à filtre, Finntip™ Filter
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
94052000 94052020 94052100 94052150 94052200 94052320 94052350 94052450 94052310 94052430 94052060 94052550 94052160 94052410 94052300 94052800
613-2656EA 154 EUR
613-2656 613-4490 613-2658 613-4492 613-2659 613-4494 613-2661 613-4497 613-4493 613-4496 613-4491 613-5011 613-5023 613-4495 613-2660 613-7087
Pointes à filtre, Finntip™ Filter
Pointes de pipettes
Integral filter made from water-repellent PE which does not swell on accidental penetration by liquid, allowing recovery of the sample. Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0,2 μl to 10 ml.
  • High quality PP with PE filter
  • Sterilised by gamma irradiation, certified RNase-, DNase-, DNA- PCR inhibitors, ATP and endotoxin-free
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Filtered Finntip pipette tips are ideal for sensitive assays and with critical samples to prevent cross-contaminations
  • Inert and non-sealing polyethylene filter
  • Tips are supplied in vacuum sealed, sterilised rack
  • 96-format volumes: 10, 20, 30, 100, 200, 300 and 1000 μl
  • Extended length tips available for volumes 100, 200 and 1000 µl allowing for better reach with specific labware consumables
  • 384-format volumes: 20 and 50 µl, Macro volumes: 5 ml and 10 ml
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
Ideal for PCR and all other amplification techniques, or any other work that involves a risk of contaminating the pipette by the aerosol effect.