Imprimer
Pasteur pipettes, PE
Fournisseur: Alpha Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LW4010 LW4053 LW4114
612-1793EA 112 EUR
612-1793 612-3751 612-3752
Pasteur pipettes, PE
Pipettes Pipettes Pasteur
These single-use transfer pipettes are unbreakable, don't require a separate bulb and, as they are manufactured from soft PE plastic, can be recycled. Sterile.
612-3751 for accessing deep vessels such as Winchesters and tissue culture flasks.
612-1793: type C, with graduations at 0,25 ml.
612-3751: type E, without graduations.
612-3752: type D, with graduations at 0,5 ml.
Cdt: 20 pipettes/bag.