Imprimer
Transfer pipettes, fine tip
Fournisseur: Alpha Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LW4243 LW4233 LW4060 LW4235 LW4239 LW4238 LW4232 LW4061 LW4236 LW4070 LW4697 LW4638 LW4636
612-1738EA 165 EUR
612-1738 612-1751 612-1759 612-1771 612-1787 612-1797 612-0006 612-0003 ALPCLW4236 ALPCLW4070 ALPCLW4697 ALPCLW4638 ALPCLW4636
Transfer pipettes, fine tip
Pipettes Pasteur/Transfer Pipettes
Non toxic, food grade LDPE.
  • For careful control and small volume transfers
  • Ideal for supernatant removal, slide preparation and gel loading