Imprimer
Serological pipettes, MBL
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PRA840S PRA842S PRA844S PRA846S PRA848S
612-2950EA 19.8 EUR
612-2950 612-2951 612-2952 612-2953 612-2954
Serological pipettes, MBL
Pipettes Pipettes sérologiques
Soda-lime glass, graduated, type 4.
  • Calibrated to deliver from any graduation line down to zero at the jet, with the last drop expelled by blowing
  • Tooled suction ends accept cotton wool plugs
  • Graduations in permanent amber stain
Certifications: ISO 835