Imprimer
Safety pipette fillers, Standard/Universal/Flip
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-1930EA 21.6 EUR
612-1930 612-2008 612-1931 612-2004 612-1920 612-1947
Safety pipette fillers, Standard/Universal/Flip
Pipettes Pipette Bulbs
Natural rubber.
  • For measuring and volumetric pipettes up to 10 ml (Standard) or 100 ml (Universal/Flip)
  • Standard/Universal: Three ventilation valves
  • Flip: Only two operating points for easier usage (release through an automatic removable valve)