Imprimer
Pipettes à poire, deux traits, classe AS
Fournisseur: HIRSCHMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1340229 1340233 1340240 1340248 1340255 1340260 1340264 1340265 1340270 1340275 1340280
612-4874EA 16.7 EUR
612-4874 612-2783 612-2784 612-2785 612-2786 612-2787 612-2788 612-2789 612-2790 612-2791 612-2792
Pipettes à poire, deux traits, classe AS
Pipettes Pipettes jaugées
AR-Glas®
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in amber stain
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
DIN 12691
Certifications: DIN EN ISO 648; DE-M marking