Imprimer
Adhesion slides, HistoBond®
Fournisseur: MARIENFELD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0810000 0810001 0830001
631-0624EA 21.1 EUR
631-0624 631-0625 630-2830
Adhesion slides, HistoBond®
Slides Adhesive Slides
The glass surface of the HistoBond® slides is loaded with a permanent positive charge which fastens all types of section to the glass. Proteinic adhesives, which often interfere with stains and immunoreactions or have disturbing background effects, are no longer required. The tissue sections adhere to the glass when subjected to frequent washing or even cooking in the microwave. In most cases, this reliable bonding makes it possible to dispense with double experiments which saves time and reduces the costs of routine section preparations. HistoBond® adhesive microscope slides have silane treated surfaces are pre-cleaned and ready for use, autoclavable with silky frosted marking area of approx. 20 mm (at one end, on both sides).
  • Ground edges 90°
  • Twin frosted 20 mm at one end, both sides
W×D×H: 76×26×1 mm