Imprimer
Microplaques, profil abaissé, pour HLA, MicroWell™, Nunc™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
438733 439225
732-2750EA 425 EUR
732-2750 732-2751
Microplaques, profil abaissé, pour HLA, MicroWell™, Nunc™
Microplaques Microplaques immunologiques et bandelettes
PS, with lid, non sterile. The low profile design and 10 µl well volume make these plates ideal for serological applications.
  • Limited adsorption of proteins
  • Optical clarity allows use with inverted microscopes
  • Lids have keyed tabs for proper orientation
  • Large raised alphanumeric coordinates
  • Unique surface treatment to maximise sample and reagent mixing
Colour: Clear.
Well dimensions (mm): Top 3,71, Bottom 1,27, Depth 1,91.
External dimensions (mm): 84×59 (with lid).