Imprimer
Mixing measuring cylinders, tall form, class B, borosilicate glass, PYREX®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3211/10MP 3211/12MP 3211/08MP 3211/16MP 3211/04MP 3211/06MP 3211/14MP 3211/18MP
612-0076EA 115 EUR
612-0076 BIBB3211/12MP 612-0075 BIBB3211/16MP BIBB3211/04MP BIBB3211/06MP BIBB3211/14MP BIBB3211/18MP
Mixing measuring cylinders, tall form, class B, borosilicate glass, PYREX®
Éprouvettes Éprouvettes graduées
Borosilicate glass, with polyethylene (PE) stopper, class B.
  • Cylinders have hexagonal foot
  • Graduations in white enamel for improved visibility with dark coloured solutions
Height values = maximum height
Certifications: ISO 4788