Imprimer
Measuring cylinders
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
34808 34820 34828 34838 34848 34854 34862 34864 35008 35020 35028 35038 35048 35054 35062 35064
612-4358EA 8.3 EUR
612-4358 612-4359 612-4360 612-4361 612-4362 612-4363 612-4364 612-4365 612-4641 612-4642 612-4643 612-4644 612-4645 612-4646 612-4647 612-4648
Measuring cylinders
Cylinders Measuring Cylinders
PP, highly translucent, tall form, class B.
  • Calibrated to contain (TC, In)
  • With blue printed scale or embossed scale
  • Exposure to temperatures up to +80 °C will not cause permanent exceeding of tolerance limits; Cleaning temperatures below +60 °C is recommended to preserve marks and inscriptions
DIN 12 681 / ISO 6706