Imprimer
Mixing measuring cylinders, SILBERBRAND ETERNA
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
33962 33908 33920 33928 33938 33948 33954
612-5044EA 154 EUR
612-5044 612-5038 612-5039 612-5040 612-5041 612-5042 612-5043
Mixing measuring cylinders, SILBERBRAND ETERNA
Cylinders Measuring Cylinders
Borosilicate glass 3.3, class B.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5043 and 612-5044 octagonal PE stopper)
  • ETERNA amber coloured line graduations
DIN EN ISO 4788
Certifications: DIN EN ISO 4788