Imprimer
VWR®, Measuring Cylinders, Tall Form, Class B, PP
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-4401EA 90.8 EUR
612-4401 612-4402 612-4403 612-4404 612-4405 612-4406 612-4407
VWR®, Measuring Cylinders, Tall Form, Class B, PP
Cylinders Measuring Cylinders
PP, translucent, tall form, class B.
  • Ring-moulded graduations, overprinted in blue
  • Large hexagonal base for better stability
  • Anti-drip lip
  • Autoclavable at 121 °C for 20 minutes
Non wetting properties mean no loss of liquid. The measured liquid quantity is the same as the quantity dispensed.
DIN 12681, ISO 6706
Certifications: DIN 12681, ISO 6706