Imprimer
Container with snap-on lid, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7142-0015 7142-0020
216-4429EA 103 EUR
216-4429 216-4430
Container with snap-on lid, Nalgene®
Barrels, Kegs, Drums
HDPE, translucent.
  • Good chemical resistance
  • Heat-resistant from −100 to +120 °C
  • Convenient handles, stackable
For storing and transporting solid and liquid materials, samples and general labware. With tight seal. Approved by the National Sanitation Foundation (NSF) for storing and transporting food and drink.