Imprimer
Dissolved oxygen probes, Orion
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
083010MD 083005MD
662-1136EA 945 EUR
662-1136 664-0152
Dissolved oxygen probes, Orion
Electrodes Dissolved Oxygen Electrodes
These standard DO probes can be used in the lab or in the field.
  • Durable and accurate
  • Sensor types: Dual thermally separated
  • Automatic temperature compensation
  • Convenient screw-on membrane caps
  • Response time: 90% of final value in 10 sec.
DO probe Orion 083005A is for use with Orion 830A, 835A meters. Cable length: 1,5 m, 8-pin waterproof DIN connection.
DO Probe Orion 083010A is for use with Orion 862A meter. Cable length: 3 m, 8-pin waterproof DIN connection.
DO Probe Orion 083005D is for use with Orion 810Aplus, 850Aplus meters. Cable length: 1,5 m, 8-pin DIN connection.
DO Probe Orion 083005MD is for use with Orion Star™ Series meters. Cable length: 1,5 m, 9-pin MiniDIN connector.
Informations sur les accessoires: These BOD adapters for polarographic dissolved oxygen probes help ensure proper and optimal probe performance. Adapters include stir funnel and electrode adapter sleeve for BOD bottle.