Imprimer
Separating funnels, Squibb
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
04010028 04010037 04010049 04010058 04010070 04010073 04012049 04012058 04014128 04014137 04014149 04014158 04014170 04014173 04012073 04012028 04012037 04012070 04016128 04016137 04016149 04016158 04016170 04016173
527-1331EA 74.3 EUR
527-1331 527-1312 527-1333 527-1334 527-1335 527-1336 527-1992 527-1993 527-1321 527-1322 527-1323 527-1324 527-1325 527-1326 527-1995 527-1990 527-1991 527-1994 527-1301 527-1302 527-1303 527-1304 527-1305 527-1306
Separating funnels, Squibb
Funnels Separating Funnels
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.