Imprimer
Vacuum filtration units
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16307 16316
516-7016EA 348 EUR
516-7016 512-3321
Vacuum filtration units
Funnels Membrane Filter Funnels
Borosilicate glass, anodised aluminium clamp, silicone lid, autoclavable.
516-7016: With glass frit filter support
512-3321: With PTFE-coated stainless steel screen filter support