Imprimer
Vacuum membrane filter holder, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1960-002 1960-004 1960-009 1960-032 1960-052 1960-054
516-0613EA 667 EUR
516-0613 516-0614 516-0615 511-0137 512-0641 516-0616
Vacuum membrane filter holder, Whatman™
Entonnoirs Entonnoirs filtrants avec membrane
Produced from borosilicate glass and available with a choice of support screen.
  • Sintered glass support is recommended for filtration and biological analysis
  • 304 stainless steel support screen is suitable for use with proteinaceous solutions
Suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
Chemical resistant to most aqueous and organic solutions as well as acid and caustic solution resistant.
Applications include: - food & beverage (e.g cold sludge in beer; ice cream), pharmaceuticals and cosmetics, water and wastewater, residue analytics and precipitation analysis, contamination tests (e.g in electroplating), microbiological, biochemical, and hydrobiological detection, radiochemical tests, particle analysis in sensitive areas of electronics, aviation, and space travel.