Imprimer
Büchner funnels, Hartley pattern
Fournisseur: DIXON SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
237091601 237091602 237091603
511-0837EA 404 EUR
511-0837 511-0838 511-0839
Büchner funnels, Hartley pattern
Entonnoirs Entonnoirs pour la filtration Entonnoirs Büchner
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.