Imprimer
3-piece filter funnels, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1950-002 1950-004 1950-007 1950-009 1950-012 1950-017 1950-027
516-0605EA 352 EUR
516-0605 516-0606 516-0607 516-0608 516-0609 516-0610 516-0611
3-piece filter funnels, Whatman™
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
Whatman 3-piece filter funnels from Cytiva are ideal for quick and easy filtration using high-efficiency glass microfiber filters.
  • Chemically resistant - 3-piece construction consists of glass cylinder and base with a polymeric membrane support
  • Support options - Provided with acrylic support and optional polypropylene or PTFE supports available separately for some sizes
  • Secure sealing - Clamps at edges to prevent peripheral loss due to fluid bypass around the edge of the filter
  • Eliminate carry-over - Simplicity of design facilitates easy and thorough cleaning between use
The increased use of high-efficiency glass microfiber filters in modern laboratories has created a demand for simple and effective filter-holding systems. Whatman 3-piece Filter Funnels have been designed to complement the range of Whatman fine particle retention, rapid flow rate glass microfiber filters.
The unit consists of a glass funnel and base, acrylic support platform, and 3 easy-to-apply metal clips. The range of diameters, funnel sizes, reservoir volumes and alternate support platforms provides a flexible system to meet the varied and unique needs of busy laboratories.