Imprimer
Dropping funnels, cylindrical, without pressure balance
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3590/02 3590/04 3590/06 3590/08 3590/01
BIBB3590/02EA 129 EUR
BIBB3590/02 BIBB3590/04 BIBB3590/06 BIBB3590/08 BIBB3590/01
Dropping funnels, cylindrical, without pressure balance
Entonnoirs Ampoules de coulée
PYREX®, borosilicate glass.
  • Interchangeable glass key
  • High chemical temperature resistance
  • Double graduated white scale showing approximate volume contained/dispensed
Cylindrical shape, with standard ground joint socket and stopper.
Certifications: ISO 4800 type 4