Imprimer
Forceps
Fournisseur: OUTILS RUBIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
K6 K35A KR K2A K7
232-1313EA 18.1 EUR
232-1313 232-1317 232-1319 232-0210 232-0204
Forceps
Forceps
Polybutene terephthalate (PBT) reinforced with 20% fibre glass, white. Universal forceps for a variety of laboratory and electronics-based applications.
  • Very good chemical resistance, anti-magnetic, high mechanical strength
  • High insulating values
  • Heat resistant up to 210 °C (for short periods)