Imprimer
Volumetric flasks, class A, white markings
Fournisseur: Poulten & Graf
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RB258/AM/A/5 RB258/AM/D/5 RB258/AM/E/5 RB258/AM/J/2 RB258/AM/H/2 RB258/AM/I/2 US258/AM/WACA5 US258/AM/WACD5 US258/AM/WACE5 US258/AM/WACF5 US258/AM/WACG5 US258/AM/WACH2 US258/AM/WACI2 US258/AM/WACJ2 US258/AM/WACK2 RB258/AM/WAC/D RB258/AM/WAC/E RB258/AM/WAC/F RB258/AM/WAC/G RB258/AM/WAC/H RB258/AM/WAC/I RB258/AM/WAC/K
612-1856EA 170 EUR
612-1856 612-1858 612-1859 612-1872 612-1870 612-1871 POULUS258/AM/WACA5 POULUS258/AM/WACD5 POULUS258/AM/WACE5 POULUS258/AM/WACF5 POULUS258/AM/WACG5 POULUS258/AM/WACH2 POULUS258/AM/WACI2 POULUS258/AM/WACJ2 POULUS258/AM/WACK2 POULRB258/AM/WAC/D POULRB258/AM/WAC/E POULRB258/AM/WAC/F POULRB258/AM/WAC/G POULRB258/AM/WAC/H POULRB258/AM/WAC/I POULRB258/AM/WAC/K
Volumetric flasks, class A, white markings
Ballons Fioles jaugées
Borosilicate glass, amber, with PP NS-stopper.
  • Easy to read white markings
  • Very good chemical resistance
  • Very good temperature resistance
Amber coloured to protect light sensitive substances.
Certifications: Acc. to ISO and ASTM or USP standards