Imprimer
Volumetric flasks, trapezoidal shape, class A, brown markings
Fournisseur: Poulten & Graf
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RB257/B/5 RB257/D/5 RB257/E/5 RB257/AM/C/5 RB257/AM/D/5
612-1848EA 130 EUR
612-1848 612-1849 612-1852 612-1839 612-1843
Volumetric flasks, trapezoidal shape, class A, brown markings
Ballons Fioles jaugées
Borosilicate glass, with PP NS-stopper.
  • Individually numbered
Individually numbered.
Certifications: DIN/BS/ISO 1042