Imprimer
Volumetric flasks, class A, white markings, works certified
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3151/04WC 3151/06WC 3151/08WC 3151/10WC 3151/12WC 3151/14WC 3151/16WC 3151/18WC 3151/20WC 3151/22WC 3151/24WC 3151/26WC 3151/01WC 3151/02WC
612-3232EA 179 EUR
612-3232 612-3239 612-3202 612-3237 612-3234 612-3220 612-3214 612-3199 612-3229 612-3227 612-3225 612-3223 612-3200 612-3201
Volumetric flasks, class A, white markings, works certified
Ballons Fioles jaugées
PYREX®, borosilicate glass, with PE stopper.
  • Calibrated to the tolerances required by the United States Pharmacopoeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with individual serial number and date of manufacture
  • Calibrated to contain (TC, In)
  • White enamel inscriptions
Complies with ISO 1042 and DIN 12664.
Informations de commande: Supplied with certificate of calibration traceable to National Standards.