Imprimer
Volumetric flasks, class A, blue graduation, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
37234 37236 37253 37254 37255 37291 37238 37243 37246 37247 37248 37251 37252 37290 37293 37249 37250 37245 37241_U 37233 37241
612-3689EA 46.2 EUR
612-3689 612-2737 612-5082 612-5083 612-3175 612-3146 612-5073 612-5074 612-5075 612-5076 612-5077 612-5080 612-5081 612-3141 612-3136 612-5078 612-5079 612-3653 612-6788 612-6787 612-6874
Volumetric flasks, class A, blue graduation, BLAUBRAND®
Flasks Volumetric Flasks
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • Conformity certified
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Informations de commande: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.