Imprimer
Flasks, round bottom, with three necks and Quickfit® ground joints
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FR250/3S/22P FR2L/3US/22P FR5L/5US/33P FR1L/3S/22P FR500/3S/22P FR1L/4S/22P FR250/4S/11P FR2L/4S/22P FR2L/4S/44P FR1L/5S/22P FR3L/5S/22P FR500/4S/22P FR1L/5S/33P FR2L/5S/33P FR3L/5S/33P
201-8507EA 90 EUR
201-8507 201-8514 201-8548 BIBBFR1L/3S/22P BIBBFR500/3S/22P BIBBFR1L/4S/22P BIBBFR250/4S/11P BIBBFR2L/4S/22P BIBBFR2L/4S/44P BIBBFR1L/5S/22P BIBBFR3L/5S/22P BIBBFR500/4S/22P BIBBFR1L/5S/33P BIBBFR2L/5S/33P BIBBFR3L/5S/33P
Flasks, round bottom, with three necks and Quickfit® ground joints
Ballons Flacons à fond rond
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Central neck and two parallel side necks. Matt white labelling area.