Imprimer
Vacuum filtering flasks, with side arm, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DS4101-1000 DS4101-2000
516-0580EA 61.4 EUR
516-0580 215-8248
Vacuum filtering flasks, with side arm, Nalgene®
Flasks Filter Flasks
PP, white.
  • Tubulation at 45° angle prevents tipping
  • Barbed adapter accepts 6,4 to 9,5 mm internal diameter tubing
  • Oval marking area
  • 215-8248 is graduated
Moulded in one piece to withstand full vacuum to 30-in. Hg.