Imprimer
Flacons de culture Erlenmeyer avec bouchons à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4112-2800 4113-2800 4115-2800 4116-2800
214-7107EA 273 EUR
214-7107 214-7108 214-7109 214-7110
Flacons de culture Erlenmeyer avec bouchons à vis, Nalgene®
Ballons Flacons Erlenmeyers
Clear PETG, with flat or baffled bottom.
  • Available with either spill-proof HDPE screw cap or vented screw cap with membrane
  • Packaged individually for easy storage and handling
  • Reduced risk of cross-contamination
Sterile, disposable culture flasks reduce the risk of cross-contamination. Ideal for shaker and suspension cultures, and for preparing or storing media. Moulded-in graduations indicate approximate contents.