Imprimer
Stirrer flasks, four arm, Celstir®, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
356984 356987
734-3190EA 1420 EUR
734-3190 734-3191
Stirrer flasks, four arm, Celstir®, WHEATON®
Flasks Spinner Flasks
Borosilicate glass.
This microcarrier spinner flask is constructed of borosilicate glass, a fluropolymer resin-faced rubber-lined cap, has an even stirring action, and is fully autoclavable.