Imprimer
Flacons à culture plats en verre clair
Fournisseur: RIXIUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10600005022OSV 10601005022OSV 1-0601-0250-28 1-0601-0250-22 1-0601-0500-28 1-0601-1000-28 1-0601-0100-22 10600010022OSV 10600025022OSV 1-0600-0050-22 1-0600-0100-22 1-0600-0250-22 1-0601005022 1-0601-010022W 1-0601-025022W 1-0601-025028W 1-0601-050028W 1-0601-100028W
RIXI10600005022OSVEA 174 EUR
RIXI10600005022OSV RIXI10601005022OSV RIXI1-0601-0250-28 RIXI1-0601-0250-22 RIXI1-0601-0500-28 RIXI1-0601-1000-28 RIXI1-0601-0100-22 215-4665 215-4666 215-4738 215-4739 215-4740 215-4741 215-4742 215-4743 215-4744 215-4745 215-4746
Flacons à culture plats en verre clair
Flacons Flacons en verre
Clear or amber glass with or without black PP cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Octagonal, flat shape with round thread in accordance with DIN 168
Ideal for applications involving cell culture. Amber coloured to protect light-sensitive materials.