Imprimer
Erlenmeyer flasks, wide neck, with flared rim
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1140/14D 1140/18D 1140/02M 1140/04D 1140/08D 1140/10D
214-0349EA 18.8 EUR
214-0349 214-0350 214-0041 214-0042 214-0043 214-0044
Erlenmeyer flasks, wide neck, with flared rim
Ballons Flacons Erlenmeyers
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With white graduations and marking spot.
Certifications: DIN 12385