Imprimer
Erlenmeyer flasks, narrow or wide mouth, with beaded rim
Fournisseur: VWR Collection
Ratings:
Moyenne des Evaluations: 5.0 (1 Avis)
Total Evaluation: 1
Moyenne des Notes : 5.0 sur 5
Cet avis vous a-t-il aidé ? Oui Non
Oui : 4 Non : 5 
aquaculture / responsable laboratoire / l'epine
Oui, je le conseille
erlens pour culture de phytoplancton, utilisés tous les jours, passer à l'acide et autoclaves. Bonne résistance
214-1130EA 129 EUR
214-1130 214-1131 214-1132 214-1133 214-1134 214-1136 214-1137 214-1170 214-1171 214-1172 214-1174 214-1175 214-1173 214-0248 214-0471 214-0472
Erlenmeyer flasks, narrow or wide mouth, with beaded rim
Flasks Erlenmeyer Flasks
Borosilicate 3.3 glass, clear.
  • Narrow or wide neck, with beaded rim
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
An economical alternative for a wide range of laboratory applications. Flasks have flared rim with markings for approximate volume graduations and a white labelling area.
ISO 1773 (narrow mouth)
DIN ISO 24450 (wide mouth)
Certifications: ISO 1773 (narrow neck), DIN ISO 24450 (wide neck).