Imprimer
Dispenser for eyewash pods
Fournisseur: SAFETY FIRST AID GROUP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E440 E401A/25
118-0116EA 0 EUR
118-0116 118-0113
Dispenser for eyewash pods
Matériel de premiers secours Stations et douches pour le lavage oculaire
Wall-mounted eyewash pod dispenser with integrated mirror for easier self-administration.
  • Clear instructions for use printed on front of cabinet
  • Dust and dirt proof
  • Will hold up to 30 pods (supplied empty)
Informations de commande: Sterile pods (118-0113) need to be ordered separately.