Imprimer
Filtres de protection sous vide, VACU-GUARD™ et VACU-GUARD™ 150, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6722-1001 6722-1002
189-0300EA 304 EUR
189-0300 189-0301
Filtres de protection sous vide, VACU-GUARD™ et VACU-GUARD™ 150, Whatman™
Filtres Systèmes de filtres en ligne
Whatman VACU-GUARD capsule filters are ideal for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.
  • Excellent for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols
  • Designed for in-line use with stepped barb connections for 10 to 12 mm ID hose
  • Available with choice of chemical trap: Activated carbon, molecular sieve, or desiccant
  • Membrane retains 99,99% of airbone particles greater than 0,1 µm
  • Features hydrophobic polytetrafluoroethylene (PTFE) membrane
The easy-to-use VACU-GUARD inline filter devices help to confine and isolate infectious materials in vacuum systems and protect your laboratory.