Imprimer
Filtres à seringue certifiés pour la préparation d'échantillons HPLC, SPARTAN™, Whatman™
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10463030 10463032 10463040 10463042 10463050 10463052 10463060 10463062 10463100 10463102 10463110 10463112
514-1226EA 426 EUR
514-1226 514-1234 514-1222 514-1231 514-1228 514-1230 514-1224 514-1233 514-1223 514-1232 514-1227 514-1157
Filtres à seringue certifiés pour la préparation d'échantillons HPLC, SPARTAN™, Whatman™
Filtres Filtres-seringue
SPARTAN syringe filters with regenerated cellulose (RC) membrane provide reproducible filtration of organic and aqueous solutions for high performance and ultra-high performance liquid chromatography (HPLC and UHPLC) sample preparation.
  • Hydrophilic membrane made of regenerated cellulose fiber exhibits low protein binding membrane
  • Disposable filter unit with PP housing
  • Excellent chemical resistance to common aqueous and organic solvents, so recommended for HPLC method sample preparation
  • Tested and certified for the absence of UV-absorbing substances at wavelengths of 210 and 254 nm with water, methanol, and acetonitrile
  • Documented batch-to-batch quality and consistency, ensuring reproducible results
  • Low dead volume Mini-tip option
  • Recyclable after use
HPLC sample preparation often requires a filtration step before samples can be used for liquid column chromatography. This step helps to clarify peaks of chemical compounds being analyzed and protects the HPLC column from large particles that might damage it.
SPARTAN regenerated cellulose filters from Cytiva's business cater to this need, because their regenerated cellulose hydrophilic membrane is resistant to common HPLC grade solvents and exhibits very low levels of extractables that could otherwise interfere with chromatographic analysis.
In addition to their use as HPLC syringe filters, SPARTAN filters can be used in capillary gel electrophoresis, protein sample ultrafiltration, UV/VIS analysis, and any application requiring a chemically resistant, low protein binding, hydrophilic membrane in a polycarbonate filter housing.
Certifications: Certified for HPLC sample prep.